Lithium bromide, 99+%, for analysis, anhydrous
Lithium bromide, 99+%, for analysis, anhydrous
Lithium bromide, 99+%, for analysis, anhydrous
Thermo Scientific Chemicals

Lithium bromide, 99+%, for analysis, anhydrous

CAS: 7550-35-8 | BrLi | 86.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
10 kg
Catalog number 199871000
Price (USD)
86.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
86.00
Each
Chemical Identifiers
CAS7550-35-8
IUPAC Namelithium(1+) bromide
Molecular FormulaBrLi
InChI KeyAMXOYNBUYSYVKV-UHFFFAOYSA-M
SMILES[Li+].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.0 %
Sulfate (SO4)=<200 ppm
Copper (Cu)=<3 ppm
Magnesium (Mg)=<20 ppm
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only