Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size
Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size
Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size
Thermo Scientific Chemicals

Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size

CAS: 7789-78-8 | CaH2 | 42.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Catalog number 199901000
Price (USD)
117.65
Online Exclusive
131.00
Save 13.35 (10%)
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
117.65
Online Exclusive
131.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS7789-78-8
IUPAC Namecalcium dihydride
Molecular FormulaCaH2
InChI KeyFAQLAUHZSGTTLN-UHFFFAOYSA-N
SMILES[CaH2]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Aluminium (Al)=<100 ppm
Assay>=92 % (Total Ca)
Iron (Fe)=<100 ppm
Particle size=<2 mm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only