PLUM™ Emergency Eyewash Station Refills
Fisher Scientific™

PLUM™ Emergency Eyewash Station Refills

Refill kits for PLUM station
Have Questions?
Catalog NumberProduct TypeIncludesCapacity (Metric)
19992050
also known as FISHERMIS-19992050-3/PK
Saline Eyewash Solution RefillThree 500 mL bottles of saline solution, form-fitting eye cup, a dust cap and instructions500 mL
Catalog number 19992050
also known as FISHERMIS-19992050-3/PK
Price (KRW)
-
Product Type:
Saline Eyewash Solution Refill
Includes:
Three 500 mL bottles of saline solution, form-fitting eye cup, a dust cap and instructions
Capacity (Metric):
500 mL

Refill kits for PLUM station

  • Bottles contain a sterile 0.9% sodium chloride solution (saline) for rinsing dust and debris, or a 4.9% sterile phosphate buffer (pH neutralizing) solution to quickly neutralize acids and alkalis
  • Bottles have clear first aid instructions, twist-open design for quick access and a built-in eye cup with dust cap
  • Use to refill eye wash stations or take to job sites as part of first aid kits
  • Bottles require no maintenance and have a three-year shelf life from date of manufacture
Specifications
Capacity (English)16 oz.
Capacity (Metric)500 mL
For Use With (Equipment)PLUM Eyewash Stations or Alone
IncludesThree 500 mL bottles of saline solution, form-fitting eye cup, a dust cap and instructions
Product TypeSaline Eyewash Solution Refill
Shelf Life3 Years
Unit SizePack of 3