Sulfur, 99.999%, (trace metal basis)
Sulfur, 99.999%, (trace metal basis)
Sulfur, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Sulfur, 99.999%, (trace metal basis)

CAS: 7704-34-9 | S | 32.06 g/mol
製品番号(カタログ番号)数量パッケージ
19993010010 gGlass bottle
製品番号(カタログ番号) 199930100
価格(JPY)
-
数量:
10 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7704-34-9
IUPAC Namesulfur
Molecular FormulaS
InChI KeyNINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES[S]
さらに表示
Appearance (Color)Yellow
Appearance (Form)Pieces or granules
Total trace metal impurities=<0.001 %
Particle size=<12 mm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only