Gibco™

MEM Rega-3

Catalog number: 19993013
Gibco™

MEM Rega-3

Catalog number: 19993013
Catalog Number
19993013
Unit Size
500 mL
Price (USD)
Price: 23.17
Your Price:
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
19993013500 mL
Price: 23.17
Your Price:
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Minimum Essential Medium (MEM) REGA-3 is a modification of the commonly used MEM. The Rega-3 modification is often used to culture HEP-G2 (human liver carcinoma) cells and occasionally other human tumor cells. We offer a variety of Gibco™ MEM modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM is modified as follows:

With Without
• Phenol Red • L-glutamine
• Gentamycin Sulfate • HEPES
• Sodium Bicarbonate

The complete formulation is available.

Gibco™ MEM, developed by Harry Eagle, was based on his earlier formulation of Basal Medium Eagle (BME). Many other modifications of MEM followed, including this Rega-3 modification. MEM Rega-3 is unique because it has a reduced sodium bicarbonate concentration, modified salt concentrations, and contains the antibiotic gentamycin sulfate. This product is made with Earle’s salts.

Product Use
For Research Use Only: Not intended for animal or human diagnostic or therapeutic use.

cGMP Manufacturing and Quality System
Gibco™ MEM is manufactured at a cGMP compliant facility located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

MEM Rega-3 contains no proteins, lipids, or growth factors. Therefore, MEM Rega-3 requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM uses a sodium bicarbonate buffer system (0.85 g ⁄ L) and therefore requires a low CO2 environment to maintain physiological pH.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Cell Type
Mammalian Cells
Concentration
1 X
Culture Type
Mammalian Cell Culture
Form
Liquid
Product Type
Minimum Essential Medium (MEM)
Shelf Life
12 Months
Sterility
Sterile-filtered
With Additives
Phenol Red, Sodium Bicarbonate, Gentamicin, Earle's Salts
Without Additives
No Glutamine, No HEPES
Green Features
Sustainable packaging
Included Antibiotics
Gentamicin
Shipping Condition
Room Temperature

Contents & Storage

Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture

Media Formulations