Ammonium chloride, 99.5%, extra pure
Ammonium chloride, 99.5%, extra pure
Ammonium chloride, 99.5%, extra pure
Thermo Scientific Chemicals

Ammonium chloride, 99.5%, extra pure

CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 199970010
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
120.65
Online Exclusive
134.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS12125-02-9
IUPAC Nameamine hydrochloride
Molecular FormulaClH4N
InChI KeyNLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILESN.Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Sulfated ash=<0.01 %
Insoluble matter=<0.005 %
pH4.5 to 5.5 (5 % at 20°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only