Ammonium chloride, 99.5%, extra pure
Ammonium chloride, 99.5%, extra pure
Ammonium chloride, 99.5%, extra pure
Thermo Scientific Chemicals

Ammonium chloride, 99.5%, extra pure

CAS: 12125-02-9 | ClH4N | 53.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 kg
5 kg
25 kg
Packaging:
View all
Fiber Drum
Catalog number 199970250
Price (USD)
-
Quantity:
25 kg
Packaging:
Fiber Drum
Request bulk or custom format
Chemical Identifiers
CAS12125-02-9
IUPAC Nameamine hydrochloride
Molecular FormulaClH4N
InChI KeyNLXLAEXVIDQMFP-UHFFFAOYSA-N
SMILESN.Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Argentometric>=99.5 %
Heavy metals (as Pb)=<5 ppm
Sulfated ash=<0.01 %
Insoluble matter=<0.005 %
pH4.5 to 5.5 (5 % at 20°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only