Ammonium bromide, 99+%, for analysis
Ammonium bromide, 99+%, for analysis
Ammonium bromide, 99+%, for analysis
Thermo Scientific Chemicals

Ammonium bromide, 99+%, for analysis

CAS: 12124-97-9 | BrH4N | 97.94 g/mol
数量:
25 g
250 g
1 kg
パッケージ:
すべて表示
Plastic bottle
製品番号(カタログ番号) 199980010
価格(JPY)
-
数量:
1 kg
パッケージ:
Plastic bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12124-97-9
IUPAC Nameammonium bromide
Molecular FormulaBrH4N
InChI KeySWLVFNYSXGMGBS-UHFFFAOYSA-N
SMILES[NH4+].[Br-]
さらに表示
pH4.5 to 6.0 (5 % solution)
Iron (Fe)=<1 ppm
Appearance (Color)White
Loss on drying=<0.2 % (105°C)
Heavy metals (as Pb)=<10 ppm
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only