EPA 8270D Analyzer Kit
Thermo Scientific™
EPA 8270D Analyzer Kit
Update your lab and ease the implementation and management of semi-volatiles (SVOC) analysis regardless of the method complexity and experience level of the user.
Have Questions?
Catalog NumberSoftwareFor Use With (Equipment)
1R120400-8270Chromeleon 7.2SR4MUB or Newer, TraceFinder EFS4.1 or NewerGC-MS System
Catalog number 1R120400-8270
Price (CLP)
-
Software:
Chromeleon 7.2SR4MUB or Newer, TraceFinder EFS4.1 or Newer
For Use With (Equipment):
GC-MS System
Get a complete solution designed to provide seamless migration to Thermo Scientific™ GC-MS instrumentation for semi-volatile organic compound (SVOC) analysis. The Thermo Scientific™ EPA 8270D analyzer kit removes the need for new method development offering a validated methodology that includes instrument and processing methods, as well an EPA environmental specific report package. All components of the kit were developed to fulfill EPA methods requirements.
The EPA 8270D Kit is designed for the Thermo Scientific™ ISQ Single Quadrupole GC-MS systems coupled with the Thermo Scientific™ TRACE™ 1300 Series Gas Chromatograph. Our innovative system will allow labs updating their current GC-MS system to take advantage of key features including modular GC injectors and detectors as well as our removable ion source under vacuum providing reliability and improved productivity to routine environmental labs.
The EPA 8270D Analyzer Kit includes:
  • Column, liners, septa and ferrules specifically designed for EPA semi-volatile analysis
  • CD containing with specific instrument and data processing methods e-workflow, compound retention time database, environmental method specific reports and an instructional user guide
  • Video tutorial for method set-up to ensure that your instrument is up and running with the EPA Method 8270D immediately following system installation
  • Based upon your preference, either the Thermo Scientific™ Dionex™ Chromeleon™ Chromatography Data System (version 7.2 SR4 MUB or newer) or the Thermo Scientific™ TraceFinder™ software (EFS version 4.1 or newer) will guarantee that you meet all reporting requirements
Specifications
Description
88 Validated Compounds
Linearity Range: 0.2-200ppm (with same column and liner)
For Use With (Equipment)GC-MS System
SoftwareChromeleon 7.2SR4MUB or Newer, TraceFinder EFS4.1 or Newer
TypeAnalyzer Kit
Unit Size-