TriPlus™ RSH SMART Autosampler
Actual product may vary
TriPlus™ RSH SMART Autosampler
TriPlus™ RSH SMART Autosampler
TriPlus™ RSH SMART Autosampler
TriPlus™ RSH SMART Autosampler
TriPlus™ RSH SMART Autosampler
TriPlus™ RSH SMART Autosampler
Thermo Scientific™
TriPlus™ RSH SMART Autosampler
Extend automation, performance, and versatility for GC and GC/MS applications, while ensuring easier and safer operations through SMART consumables ID recognition and usage tracking.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct Type
1R77010-2003Advanced Liquid, Regular Rail
1R77010-2001Standard Liquid, Regular Rail
1R77010-2002Standard Liquid and Headspace, Regular Rail
1R77010-2004Advanced Liquid, Extended Rail
Catalog number 1R77010-2003
Price (MXN)
-
Product Type:
Advanced Liquid, Regular Rail
Expand automated sampling capabilities beyond liquid, headspace, and solid phase microextraction (SPME) injections. The advanced, built-in robotics of the Thermo Scientific™ TriPlus™ RSH SMART Autosampler deliver exceptional precision, sample handling flexibility, and reliable unattended operation. Enabling SMART consumables tracking, it offers easier, safer and traceable operations. Accurate and repeatable sample preparation and injection are essential to achieving the highest quality GC/GC-MS results. Enhance data quality and laboratory efficiency by automating your entire sample handling workflow.
The TriPlus RSH SMART Autosampler brings together technological innovations that further expand automation, performance and versatility for GC and GC-MS applications, while ensuring easier and safer operations through SMART consumables ID recognition and usage tracking. Automated high-precision sample handling eliminates the manual labor and downtime typically required for tasks as standard/sample dilutions, internal standard addition and derivatization.
SMART Consumables
Validated SMART consumables are equipped with unique SMART chip storing parameters, ranges and usage history; enabling full traceability of sample preparation down to the syringe.
Automatic Syringe Changeout
Automatic Tool Change (ATC) capability automates the use of different syringes for sample preparation steps or different injection techniques within the sequence.
Vial Bottom Sensing Expands Application Range
Precise detection of vial bottoms enables the autosampler to accurately withdraw from sample amounts as little as 5 μL, enhancing data quality of sample-limited applications.
Flexible liquid sample handling
Optimized liquid injection modes and customizable syringe filling techniques, support a wide range of sample types and full GC inlet compatibility.
Automated headspace injection for volatile analysis
High temperature gas-tight syringe allows direct headspace injection into the GC inlet for increased injection volume flexibility. Sample vial capacity of up to 300x 10 mL/20 mL vials and sample incubation overlapping, facilitate long unattended operation.
Automated, solvent-free SPME
Precise control of all steps—fiber preconditioning, adsorption and desorption, as well as fiber washes—delivers maximum performance for SPME applications. Boost productivity with the optimized SPME Arrow design. It offers optimized fiber geometry to achieve 10x superior sensitivity compared to classical SPME, 2x faster extraction time and superior robustness.
Automated Sample Preparation
The Triplus RSH SMART Autosampler combines highly versatile sample injection capability with automated sample preparation procedures, eliminating typical GC-MS workflow bottlenecks. Daily operations ranging from sample/standard dilution to more complex derivatization protocols, can be fully automated and easily programmed with the Sampling Workflow Editor Software.
Extend productivity with the optional 1D barcode reader
The optional 1D barcode reader eliminates the need to log and track sample information manually; enabling you to streamline internal operations, save time, increase efficiency, and reduce errors.
Specifications
DescriptionConfigured for liquid injection on regular X-axis length. Complete with liquid syringe tool, 2 x 10 μL SMART syringes, a standard washing station and a single tray holder with 3 trays for 2 mL vials. Expandable to other sampling techniques (Headspace, SPME, SPME Arrow, ITEX-DHS) and with sample preparation tools.
QuantityEach
Tool ExchangeATC or Tool Release Station
For Use With (Equipment)Compatible with Smart syringes/fibers only.
Injection Mode8 Fully Customizable Method-Specific Injection Mode Presets
Injection SpeedSelectable from 0.1 μL/s to 2000 μL/s
Injection Volume0.1 to 10,000 μL
Product TypeAdvanced Liquid, Regular Rail
Sample Capacity4608 Well Plates, 840 x 0.5 mL or 0.7 mL Vials, 648 x 2 mL Sample Vials, 240 x 10 mL or 20 mL Vials
Unit SizeEach