Tungsten(VI) chloride, 99%, 20 mesh
Tungsten(VI) chloride, 99%, 20 mesh
Tungsten(VI) chloride, 99%, 20 mesh
Thermo Scientific Chemicals

Tungsten(VI) chloride, 99%, 20 mesh

CAS: 13283-01-7 | Cl6W | 396.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 200300100
Price (TWD)
-
Quantity:
10 g
Request bulk or custom format
Chemical Identifiers
CAS13283-01-7
IUPAC Nametungsten hexachloride
Molecular FormulaCl6H6W
InChI KeyXHYHOBJDYHRWHO-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.Cl.Cl.[W]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey to dark blue to violet to black
Appearance (Form)Powder
Purity>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only