Cloruro de tungsteno(VI), 99 %, malla 20, Thermo Scientific Chemicals
Cloruro de tungsteno(VI), 99 %, malla 20, Thermo Scientific Chemicals
Cloruro de tungsteno(VI), 99 %, malla 20, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cloruro de tungsteno(VI), 99 %, malla 20, Thermo Scientific Chemicals

CAS: 13283-01-7 | Cl6W | 396.57 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
50 g
250 g
Número de catálogo 200300500
Precio (USD)
-
Cantidad:
50 g
Pedido a granel o personalizado
Identificadores químicos
CAS13283-01-7
IUPAC Nametungsten hexachloride
Molecular FormulaCl6H6W
InChI KeyXHYHOBJDYHRWHO-UHFFFAOYSA-N
SMILESCl.Cl.Cl.Cl.Cl.Cl.[W]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Grey to dark blue to violet to black
Appearance (Form)Powder
Purity>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only