(-)-Diethyl D-tartrate, 99%, made from unnatural tartaric acid
(-)-Diethyl D-tartrate, 99%, made from unnatural tartaric acid
(-)-Diethyl D-tartrate, 99%, made from unnatural tartaric acid
Thermo Scientific Chemicals

(-)-Diethyl D-tartrate, 99%, made from unnatural tartaric acid

CAS: 13811-71-7 | C8H14O6 | 206.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Catalog number 200345000
Price (USD)
-
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13811-71-7
IUPAC Name1,4-diethyl (2S,3S)-2,3-dihydroxybutanedioate
Molecular FormulaC8H14O6
InChI KeyYSAVZVORKRDODB-WDSKDSINSA-N
SMILESCCOC(=O)[C@@H](O)[C@H](O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4450 to 1.4470 (20°C, 589 nm)
Specific optical rotation-7.5° to -9.5° (20°C, 589 nm) (neat)
Appearance (Color)Clear colorless or light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only