4-Iodopyrazole, 98%
4-Iodopyrazole, 98%
4-Iodopyrazole, 98%
Thermo Scientific Chemicals

4-Iodopyrazole, 98%

CAS: 3469-69-0 | C3H3IN2 | 193.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 200370050
Price (USD)
109.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
109.00
Each
Chemical Identifiers
CAS3469-69-0
IUPAC Name4-iodo-1H-pyrazole
Molecular FormulaC3H3IN2
InChI KeyLLNQWPTUJJYTTE-UHFFFAOYSA-N
SMILESIC1=CNN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point107°C to 112°C
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Appearance (Color)White to off-white
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only