2,2,6,6-Tetramethylpiperidinooxy, 98%
2,2,6,6-Tetramethylpiperidinooxy, 98%
2,2,6,6-Tetramethylpiperidinooxy, 98%
Thermo Scientific Chemicals

2,2,6,6-Tetramethylpiperidinooxy, 98%

CAS: 2564-83-2 | C9H18NO | 156.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 200380050
Price (EUR)
43,20
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
43,20
Each
Chemical Identifiers
CAS2564-83-2
IUPAC Name(2,2,6,6-tetramethylpiperidin-1-yl)oxidanyl
Molecular FormulaC9H18NO
InChI KeyQYTDEUPAUMOIOP-UHFFFAOYSA-N
SMILESCC1(C)CCCC(C)(C)N1[O]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or solid and/or
Infrared spectrumConforms
Appearance (Color)Orange to red
Appearance (Form)chunks
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only