4-Fluoro-3-nitrobenzotrifluoride, 96%
4-Fluoro-3-nitrobenzotrifluoride, 96%
4-Fluoro-3-nitrobenzotrifluoride, 96%
Thermo Scientific Chemicals

4-Fluoro-3-nitrobenzotrifluoride, 96%

CAS: 367-86-2 | C7H3F4NO2 | 209.1 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
Packaging:
Glass bottle
Catalog number 200610050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS367-86-2
IUPAC Name1-fluoro-2-nitro-4-(trifluoromethyl)benzene
Molecular FormulaC7H3F4NO2
InChI KeyHLDFCCHSOZWKAA-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC(=CC=C1F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow
GC>=95 %
Refractive index1.4610 to 1.4630 (20°C, 589 nm)
Appearance (Form)Liquid
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only