Phosphorus pentoxide, 98%, extra pure
Phosphorus pentoxide, 98%, extra pure
Phosphorus pentoxide, 98%, extra pure
Thermo Scientific Chemicals

Phosphorus pentoxide, 98%, extra pure

CAS: 1314-56-3 | O5P2 | 141.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Glass bottle
Catalog number 200890010
Price (USD)
129.65
Online exclusive
144.00
Save 14.35
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
129.65
Online exclusive
144.00
Save 14.35
Each
Chemical Identifiers
CAS1314-56-3
IUPAC Nametricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
Molecular FormulaO5P2
InChI KeyDLYUQMMRRRQYAE-UHFFFAOYSA-N
SMILESO=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
View more
SpecificationsSpecification SheetSpecification Sheet
Reducing substance (P2O3)=<0.1 %
Appearance (Color)White
Titration with NaOH>=97.5 %
Appearance (Form)Hygroscopic crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only