Sulfur, 99.5+%, refined
Sulfur, 99.5+%, refined
Sulfur, 99.5+%, refined
Thermo Scientific Chemicals

Sulfur, 99.5+%, refined

CAS: 7704-34-9 | S | 32.06 g/mol
数量:
500 g
2.5 kg
10 kg
パッケージ:
すべて表示
Plastic drum
製品番号(カタログ番号) 201220100
価格(JPY)
-
数量:
10 kg
パッケージ:
Plastic drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7704-34-9
IUPAC Namesulfur
Molecular FormulaS
InChI KeyNINIDFKCEFEMDL-UHFFFAOYSA-N
SMILES[S]
さらに表示
Water=<0.5 %
Sulfated ash=<0.08 %
Acidity=<0.01 % (as H2SO4)
Assay>=99.50 %
Appearance (Color)Yellow
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only