Rubidium chloride, 99+%
Rubidium chloride, 99+%
Rubidium chloride, 99+%
Thermo Scientific Chemicals

Rubidium chloride, 99+%

CAS: 7791-11-9 | ClRb | 120.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 201270250
Price (EUR)
192,65
Online exclusive
214,00
Save 21,35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
192,65
Online exclusive
214,00
Save 21,35
Each
Chemical Identifiers
CAS7791-11-9
IUPAC Namerubidium(1+) chloride
Molecular FormulaClRb
InChI KeyFGDZQCVHDSGLHJ-UHFFFAOYSA-M
SMILES[Cl-].[Rb+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder or crystals and/or chunks
Total trace metal impurities=<1.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only