D(-)-4-Hydroxyphenylglycine, 98+%
D(-)-4-Hydroxyphenylglycine, 98+%
D(-)-4-Hydroxyphenylglycine, 98+%
Thermo Scientific Chemicals

D(-)-4-Hydroxyphenylglycine, 98+%

CAS: 22818-40-2 | C8H9NO3 | 167.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 201320250
Price (USD)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS22818-40-2
IUPAC Name(2R)-2-amino-2-(4-hydroxyphenyl)acetic acid
Molecular FormulaC8H9NO3
InChI KeyLJCWONGJFPCTTL-SSDOTTSWSA-N
SMILESN[C@@H](C(O)=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Specific optical rotation-155° to -161° (20°C, 589 nm) (c=1, 1N HCl)
Appearance (Form)Powder
Infrared spectrumConforms
Titration with HClO4>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only