Thermo Scientific Chemicals

Ammonium cerium(IV) nitrate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 201440050
5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium cerium(IV) nitrate, 99%, for analysis, Thermo Scientific Chemicals

Catalog number: 201440050
5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 201440050
Price (USD)
Price: 1,344.00
Online price: 1,142.65
Your price:
Quantity
-

Chemical Identifiers

CAS
16774-21-3
IUPAC Name
λ⁴-cerium(4+) bis(nitric acid) diamine tetranitrate
Molecular Formula
CeH8N8O18
InChI Key
WIBGOERAEYJBOT-UHFFFAOYSA-N
SMILES
N.N.[Ce+4].O[N+]([O-])=O.O[N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Appearance (Color)
Orange
Appearance (Form)
Crystalline powder or crystals
Titration
>=98.5 % (on dry substance)
Iron(III)oxide (Fe2O3)
=<5 ppm
Calcium oxide (CaO)
=<5 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent for analysis
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text