Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Thermo Scientific™

Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure

Conserve space with these square, narrow-mouth HDPE bottles that are chemically and break resistant, as well as economically priced for a wide range of lab uses.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호용량(미터법)마개 크기제품 사이즈
2018-003030 mL20 mmCase of 72
2018-0030PK
2018-0030으로도 사용됨
30 mL20 mmPack of 12
2018-006060 mL24 mmCase of 72
2018-0060PK
2018-0060으로도 사용됨
60 mL24 mmPack of 12
2018-0125125 mL38-430Case of 72
2018-0125PK
2018-0125으로도 사용됨
125 mL38-430Pack of 12
2018-0250250 mL38-430Case of 72
2018-0250PK
2018-0250으로도 사용됨
250 mL38-430Pack of 12
2018-10001 L38-430Case of 24
2018-1000PK
2018-1000으로도 사용됨
1 L38-430Pack of 6
카탈로그 번호 2018-0030
제품 가격(KRW)
116,000
Case of 72
용량(미터법):
30 mL
마개 크기:
20 mm
Unit Size:
Case of 72
제품 가격(KRW)
116,000
Case of 72

Conserve space with Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure. HDPE bottles are the most chemically and break resistant of all Nalgene square bottles, yet are economically priced for a wide range of lab uses. They are made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.

Features:

  • Square sides allow for good use of space and an easy grip
  • Caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Graduated for easy dispensing
  • Leakproof† when used with Nalgene closures
  • Can be used to -100°C (-148°F)
  • Not autoclavable
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Includes:

Leakproof† Polypropylene Screw Caps

Note:

Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.

Recycling Information:

Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
사양
ACT LabelYes
오토클레이브 가능Not Autoclavable
용량1 oz., 30 mL
용량(영국식 단위)1 oz.
인증/적합성USP Class VI, FDA - 21 CFR
마개 색상Natural, translucent
마개 재질Polypropylene
마개 크기20 mm
마개 유형Screw Cap
설명Narrow-Mouth Square Bottle
직경(영국식 단위)1.5 in.
직경(영국식 단위) 내부 넥0.5 in.
직경(미터법)38 mm
직경(미터법) 내부 넥14 mm
눈금Yes
높이(영국식 단위)2.5 in.
높이(미터법)64 mm
포함Leakproof polypropylene screw closures
케이스당 수72
팩당 수12
제품 유형Square Narrow Mouth Bottle
형상Square
마개 디스펜싱을 통해No
폭(영국식 단위)1.52 in.
용량(미터법)30 mL
Closure IncludedYes
색상Translucent
GraduatedYes
물질HDPE
마우스Narrow
멸균Non-sterile
Unit SizeCase of 72