Thermo Scientific Chemicals

Ammonium dihydrogen phosphate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 201830025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Ammonium dihydrogen phosphate, 99+%, for analysis, Thermo Scientific Chemicals

Catalog number: 201830025
2.5 kg, Plastic bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
7722-76-1
IUPAC Name
phosphoric acid amine
Molecular Formula
H6NO4P
InChI Key
LFVGISIMTYGQHF-UHFFFAOYSA-N
SMILES
N.OP(O)(O)=O
Appearance (Form)
Crystalline solid
Sodium (Na)
=<50 ppm
Heavy metals (as Pb)
=<5 ppm
Chloride (Cl)
=<5 ppm
Sulfate (SO4)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text