Ammonium dihydrogen phosphate, 99+%, for analysis
Ammonium dihydrogen phosphate, 99+%, for analysis
Ammonium dihydrogen phosphate, 99+%, for analysis
Ammonium dihydrogen phosphate, 99+%, for analysis
Thermo Scientific Chemicals

Ammonium dihydrogen phosphate, 99+%, for analysis

CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic bottle
Catalog number 201832500
Price (USD)
79.10
Each
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
79.10
Each
Add to cart
Chemical Identifiers
CAS7722-76-1
IUPAC Namephosphoric acid amine
Molecular FormulaH6NO4P
InChI KeyLFVGISIMTYGQHF-UHFFFAOYSA-N
SMILESN.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline solid
Sodium (Na)=<50 ppm
Heavy metals (as Pb)=<5 ppm
Chloride (Cl)=<5 ppm
Sulfate (SO4)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only