Ammonium fluoride, 98+%, extra pure
Ammonium fluoride, 98+%, extra pure
Ammonium fluoride, 98+%, extra pure
Thermo Scientific Chemicals

Ammonium fluoride, 98+%, extra pure

CAS: 12125-01-8 | FH4N | 37.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic Drum
Catalog number 201840025
Price (USD)
255.65
Special offer
Online exclusive
Ends: 28-Dec-2026
300.00
Save 44.35
Each
Quantity:
2.5 kg
Packaging:
Plastic Drum
Request bulk or custom format
Price (USD)
255.65
Special offer
Online exclusive
Ends: 28-Dec-2026
300.00
Save 44.35
Each
Chemical Identifiers
CAS12125-01-8
IUPAC Nameamine hydrofluoride
Molecular FormulaFH4N
InChI KeyLDDQLRUQCUTJBB-UHFFFAOYSA-N
SMILESN.F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder
Heavy metals (as Pb)=<5 ppm
Titration excess NaOH>=98.0 %
Residue after ignition=<0.01 % (SO4)
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only