Potassium persulfate, 99+%, for analysis
Potassium persulfate, 99+%, for analysis
Potassium persulfate, 99+%, for analysis
Thermo Scientific Chemicals

Potassium persulfate, 99+%, for analysis

CAS: 7727-21-1 | K2O8S2 | 270.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 202010250
Price (USD)
26.90
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
26.90
Each
Chemical Identifiers
CAS7727-21-1
IUPAC Namedipotassium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaK2O8S2
InChI KeyUSHAGKDGDHPEEY-UHFFFAOYSA-L
SMILES[K+].[K+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Titration Iodometric>=99.0 %
Chloride (Cl)=<120 ppm
Heavy metals (as Pb)=<50 ppm
Iron (Fe)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only