Zinc bromide, 98%, extra pure, anhydrous
Zinc bromide, 98%, extra pure, anhydrous
Zinc bromide, 98%, extra pure, anhydrous
Thermo Scientific Chemicals

Zinc bromide, 98%, extra pure, anhydrous

CAS: 7699-45-8 | Br2Zn | 225.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 202105000
Price (USD)
190.65
Online Exclusive
212.00
Save 21.35 (10%)
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
190.65
Online Exclusive
212.00
Save 21.35 (10%)
Each
Chemical Identifiers
CAS7699-45-8
IUPAC Namezinc(2+) dibromide
Molecular FormulaBr2Zn
InChI KeyVNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES[Zn++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Barium (Ba)=<10 ppm
Iron (Fe)=<10 ppm
Potassium (K)=<100 ppm
Appearance (Form)Powder and/or chunks
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only