Amberlyst™ 15, (dry) ion-exchange resin
Amberlyst™ 15, (dry) ion-exchange resin
Amberlyst™ 15, (dry) ion-exchange resin
Thermo Scientific Chemicals

Amberlyst™ 15, (dry) ion-exchange resin

CAS: 9037-24-5 | MFCD00145841
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 202145000
Price (EUR)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9037-24-5
Molecular FormulaMFCD00145841
SpecificationsSpecification SheetSpecification Sheet
Total weight capacity>=4.70 eq/kg (H)
Appearance (Form)Beads
Water=<1.5 %
Particle size=<1.0 % (< 0.300 mm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ion exchange resin
RUO – Research Use Only