Pierce™ Monomeric Avidin Agarose

Catalog number: 20228B

Thermo Scientific™  Related applications: Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Monomeric Avidin Agarose is ideal for purifying biotinylated proteins, peptides and other molecules.

Features of Monomeric Avidin Agarose:

Non-denaturing—purifies biotinylated products using mild elution conditions (2 mM free biotin)
Reusable—Monomeric Avidin Agarose can be regenerated and reused at least 10 times, with only a marginal loss in biotin binding capacity (approximately 2.5% decrease per regeneration)
Specific—retains biotin-binding specificity and low nonspecific binding of native avidin
Flexible—bind samples in a variety of physiological buffers and elute either with 0.1M glycine or by competition with 2 mM biotin
Good binding capacity—greater than 1.2 mg biotinylated BSA/mL resin

The Immobilized Monomeric Avidin protein binds biotin with high specificity and moderate affinity, allowing non-biotin molecules to be washed away and then the bound biotin-labeled molecules to be competitively eluted using 2 mM biotin in phosphate buffered saline (PBS). This technique provides the gentlest elution conditions for biotinylated protein purification and avoids the contamination and other problems associated with traditional avidin and streptavidin methods. The monomeric avidin column can then be regenerated by using 0.1M glycine to strip the column of residual bound biotin without losing the ability to bind another biotinylated sample.

With typical avidin or streptavidin, the biotin-binding affinity (Kd = 10-15M) is so great that purification with these traditional media require denaturing conditions for elution, such as 8M Guanidine·HCl at pH 1.5 or boiling in reducing SDS-PAGE sample loading buffer. In addition to their adverse effects on the biotinylated protein of interest, such harsh elution conditions typically cause extraneous proteins (i.e., the avidin or streptavidin subunits) to strip from the resin and co-elute with the desired purification product. By contrast, when avidin is coupled to a resin (e.g., crosslinked beaded agarose or UltraLink Resin) as the subunit monomer, its specificity for biotin is retained, but its biotin-binding affinity decreases to a level (Kd = 10-8M) that is conducive to subsequent elution.

Related Products
Pierce™ Monomeric Avidin Agarose Kit, 2 mL
Pierce™ Monomeric Avidin UltraLink™ Resin
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Column Type: Affinity, Agarose Resin
Format: Bottle(s)
Immobilized Phase: Avidin
Product Line: Pierce™
Product Size: Custom

Contents & storage

Upon receipt store product at 4°C. Product is shipped at ambient temperature.

Documents

Manuals & protocols