Ammonium hexafluorophosphate, 99%, pure
Ammonium hexafluorophosphate, 99%, pure
Ammonium hexafluorophosphate, 99%, pure
Acros Organics

Ammonium hexafluorophosphate, 99%, pure

CAS: 16941-11-0 | H4F6NP | 163 g/mol
Have Questions?
Catalog NumberQuantityPackaging
20232025025 gPlastic bottle
Catalog number 202320250
HSN 28352990
Price (INR)
12,799.00
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
12,799.00
Each
Chemical Identifiers
CAS16941-11-0
IUPAC Nameammonium hexafluoro-λ⁵-phosphanuide
Molecular FormulaF6H4NP
InChI KeyNIZXKAYXSNUDOU-UHFFFAOYSA-O
SMILES[NH4+].F[P-](F)(F)(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White
Elemental Analysis>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only