Bromuro de magnesio hexahidrato, +99 %, para análisis, Thermo Scientific Chemicals
Bromuro de magnesio hexahidrato, +99 %, para análisis, Thermo Scientific Chemicals
Bromuro de magnesio hexahidrato, +99 %, para análisis, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromuro de magnesio hexahidrato, +99 %, para análisis, Thermo Scientific Chemicals

CAS: 13446-53-2 | Br2H12MgO6 | 292.20 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
250 g
1 kg
Envase:
Frasco de vidrio
Número de catálogo 202510010
Precio (USD)
-
Cantidad:
1 kg
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS13446-53-2
IUPAC Namedibromomagnesium hexahydrate
Molecular FormulaBr2H12MgO6
InChI KeyLGLXXNHIGIJYQQ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.Br[Mg]Br
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Crystalline powder
pH6 to 8 (5 % at 20°C)
Sulfate (SO4)=<200 ppm
Copper (Cu)=<50 ppm
Iron (Fe)=<50 ppm
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only