Magnesium bromide hexahydrate, 99+%, for analysis
Magnesium bromide hexahydrate, 99+%, for analysis
Magnesium bromide hexahydrate, 99+%, for analysis
Thermo Scientific Chemicals

Magnesium bromide hexahydrate, 99+%, for analysis

CAS: 13446-53-2 | Br2H12MgO6 | 292.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 202512500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13446-53-2
IUPAC Namedibromomagnesium hexahydrate
Molecular FormulaBr2H12MgO6
InChI KeyLGLXXNHIGIJYQQ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.Br[Mg]Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
pH6 to 8 (5 % at 20°C)
Sulfate (SO4)=<200 ppm
Copper (Cu)=<50 ppm
Iron (Fe)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only