1,3-Bis[tris(hydroxymethyl)amino]propane, 99%
1,3-Bis[tris(hydroxymethyl)amino]propane, 99%
1,3-Bis[tris(hydroxymethyl)amino]propane, 99%
Thermo Scientific Chemicals

1,3-Bis[tris(hydroxymethyl)amino]propane, 99%

CAS: 64431-96-5 | C11H26N2O6 | 282.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
250 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number 202640010
Price (USD)
1,286.65
Online exclusive
1,429.00
Save 142.35
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
1,286.65
Online exclusive
1,429.00
Save 142.35
Each
Chemical Identifiers
CAS64431-96-5
IUPAC Name2-[(3-{[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino}propyl)amino]-2-(hydroxymethyl)propane-1,3-diol
Molecular FormulaC11H26N2O6
InChI KeyHHKZCCWKTZRCCL-UHFFFAOYSA-N
SMILESOCC(CO)(CO)NCCCNC(CO)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO498.5 to 101.5 %
UV(0.1 M in water)
UVat 400 nm A: =<0.05
Appearance (Form)Crystalline powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only