Hexamethyldisilane, 98+%
Hexamethyldisilane, 98+%
Hexamethyldisilane, 98+%
Thermo Scientific Chemicals

Hexamethyldisilane, 98+%

CAS: 1450-14-2 | C6H18Si2 | 146.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Catalog number 202690500
Price (USD)
-
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1450-14-2
IUPAC Namehexamethyldisilane
Molecular FormulaC6H18Si2
InChI KeyNEXSMEBSBIABKL-UHFFFAOYSA-N
SMILESC[Si](C)(C)[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.4211 to 1.4231 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only