Thermo Scientific™
Hemacytometer Cover Glasses
 Related applications:
Anatomical Pathology
Use alkali-free Thermo Scientific™ Hemacytometer Cover Glasses with red blood cell and platelet counting chambers.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • Precision cut
  • 0.016 in. (0.4mm) thickness
  • Two shapes available
Documents
Manuals & protocols