Sodium thiosulfate pentahydrate, 99.5%, for analysis
Sodium thiosulfate pentahydrate, 99.5%, for analysis
Sodium thiosulfate pentahydrate, 99.5%, for analysis
Thermo Scientific Chemicals

Sodium thiosulfate pentahydrate, 99.5%, for analysis

Reduction reagent | CAS: 10102-17-7 | Na2O3S2·5H2O | 248.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
2.5 kg
10 kg
Packaging:
View all
Plastic Bottle
Catalog number 202850010
Price (USD)
67.90
Each
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
67.90
Each
Chemical Identifiers
CAS10102-17-7
IUPAC Namedisodium pentahydrate sulfanidesulfonate
Molecular FormulaH10Na2O8S2
InChI KeyPODWXQQNRWNDGD-UHFFFAOYSA-L
SMILESO.O.O.O.O.[Na+].[Na+].[O-]S([S-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
SO4 and SO3 (as SO4)=<0.1 %
Insoluble matter=<0.005 % (in water)
Nitrogen compounds (as N)=<0.002 %
Sulfur (S)=<1 ppm
Chloride (Cl)=<200 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent for analysis
RUO – Research Use Only