Sodium iodide dihydrate, 99+%, for analysis
Sodium iodide dihydrate, 99+%, for analysis
Sodium iodide dihydrate, 99+%, for analysis
Thermo Scientific Chemicals

Sodium iodide dihydrate, 99+%, for analysis

CAS: 13517-06-1 | INa·2H2O | 185.93 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
產品號碼 202895000
價格 (TWD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS13517-06-1
IUPAC Namesodium dihydrate iodide
Molecular FormulaH4INaO2
InChI KeyVEEIYHUOLCUPSA-UHFFFAOYSA-M
SMILESO.O.[Na+].[I-]
檢視更多
規格Specification Sheet規格表
Titration Argentometric>=99.0 %
Iodate (IO3)=<3 ppm
Iron (Fe)=<5 ppm
pH6 to 9 (5 % in water)
Chloride and Bromide=<0.02 % (as Cl)
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only