Manganese(IV) oxide, 88%, electrolytically precipitated, active
Manganese(IV) oxide, 88%, electrolytically precipitated, active
Manganese(IV) oxide, 88%, electrolytically precipitated, active
Thermo Scientific Chemicals

Manganese(IV) oxide, 88%, electrolytically precipitated, active

CAS: 1313-13-9 | MnO2 | 86.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 203195000
Price (EUR)
94,80
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
94,80
Each
Chemical Identifiers
CAS1313-13-9
IUPAC Namedioxomanganese
Molecular FormulaMnO2
InChI KeyNUJOXMJBOLGQSY-UHFFFAOYSA-N
SMILESO=[Mn]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on drying=<3.5 %
Appearance (Color)Black
Oxidation activity>=60 % yield
Titration Iodometric>=83 % (on dried substance)
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only