Nickel(II) bromide, 99%
Nickel(II) bromide, 99%
Nickel(II) bromide, 99%
Thermo Scientific Chemicals

Nickel(II) bromide, 99%

CAS: 13462-88-9 | Br2Ni | 218.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 203360100
Price (EUR)
37,30
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
37,30
Each
Chemical Identifiers
CAS13462-88-9
IUPAC Namenickel(2+) dibromide
Molecular FormulaBr2Ni
InChI KeyIPLJNQFXJUCRNH-UHFFFAOYSA-L
SMILES[Ni++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
Water=<6 % (K.F.)
Appearance (Form)Crystalline powder and/or chunks
Titration Argentometric>=98.5 % (on dry substance)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only