Immobilized Papain (Agarose Resin)
Thermo Scientific™

Immobilized Papain (Agarose Resin)

Thermo Scientific Pierce Immobilized Papain consists of a cysteine-endopeptidase (papain) that has been immobilized onto beaded agarose resin to enableRead more
Have Questions?
Catalog NumberQuantity
203415 mL
Catalog number 20341
Price (USD)
316.65
Online exclusive
348.00
Save 31.35
Each
Quantity:
5 mL
Request bulk or custom format
Price (USD)
316.65
Online exclusive
348.00
Save 31.35
Each
Thermo Scientific Pierce Immobilized Papain consists of a cysteine-endopeptidase (papain) that has been immobilized onto beaded agarose resin to enable generation and purification of Fab fragments from antibodies.

Papain is a nonspecific, cysteine-endopeptidase isolated from papaya latex that is used in a wide variety of applications. When IgG molecules are incubated with papain in the presence of cysteine, one or more peptide bonds in the hinge region are split, producing three fragments of similar size: two Fab fragments and one Fc fragment. The Fc fragments can be separated from the Fab fragments using immobilized Protein A or by ion exchange chromatography.

The 50 kilodalton Fc fragment is often used for determining specificity of the Fc receptors without interference from antigen binding. Fab fragments are useful in immunohistochemical studies because the fragments penetrate the tissue better than intact IgG. Also, nonspecific binding from Fc receptors is avoided by using only the Fab portion of the immunoglobulin.

Papain can also can be used to generate F(ab')2 fragments. To prepare F(ab')2 fragments, the papain is first activated with 10 mM cysteine, which is then removed via gel filtration. If no cysteine is present during the papain digestion, F(ab')2 fragments can be generated. These fragments are often inconsistent and reproducibility is a problem.

Immobilized papain can be substituted for free papain in almost any application, and is advantageous because it virtually eliminates autolysis, eliminates contamination of a sample with the protease and allows control of the digestion by removing the papain or controlling the flow of sample over an immobilized papain column. Immobilized papain is also more stable against heat-induced denaturation, resulting in longer maintenance of activity. The ImmunoPure Fab Preparation Kit contains immobilized papain, other necessary reagents and an optimized protocol for human, rabbit and mouse antibody digestion.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid Suspension
Quantity5 mL
Product TypePapain
Unit SizeEach
Contents & Storage
Upon receipt store at 4°C.

Frequently asked questions (FAQs)

Can I purchase the immobilized enzymes (proteases) from the Thermo Scientific IgG Fragmentation kit separately?

Yes. The three immobilized enzymes are available in 5 mL package sizes as shown below:

• Immobilized Ficin, Cat. No. 44881
• Immobilized Papain, Cat. No. 20341
• Immobilized Pepsin, Cat. No. 20343

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

How much of the immobilized Papain (Agarose Resin) do I need for digestion of 10 mg IgG?

0.5 mL of the 50% Immobilized Papain slurry is sufficient for the digestion of up to 10 mg of pure lyophilized IgG reconstituted in 1 mL of Digestion Buffer.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.