Pierce™ High Capacity Streptavidin Agarose - FAQs
View additional product information for Pierce™ High Capacity Streptavidin Agarose - FAQs (20361B)
1 product FAQs found
What is the binding capacity of the High Capacity Streptavidin Agarose?
The High Capacity Streptavidin Agarose resin has a binding capacity of >100 µg biotinylated p-NPE/mL of settled resin and >10 mg biotinylated BSA/mL of settled resin.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.