(+)-Dihydrocarvone, 98%, mixture of isomers
(+)-Dihydrocarvone, 98%, mixture of isomers
(+)-Dihydrocarvone, 98%, mixture of isomers
Thermo Scientific Chemicals

(+)-Dihydrocarvone, 98%, mixture of isomers

CAS: 5524-05-0 | C10H16O | 152.24 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
產品號碼 203651000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS5524-05-0
IUPAC Name(2R,5R)-2-methyl-5-(prop-1-en-2-yl)cyclohexan-1-one
Molecular FormulaC10H16O
InChI KeyAZOCECCLWFDTAP-RKDXNWHRSA-N
SMILESC[C@@H]1CC[C@H](CC1=O)C(C)=C
檢視更多
規格Specification Sheet規格表
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Refractive index1.4700 to 1.4740 (20°C, 589 nm)
Specific gravity(25°C) 0.920 to 0.940
Appearance (Form)Liquid
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only