alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol
alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol
alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol
Thermo Scientific Chemicals

alpha-Terpineol, 95%, mixture of alpha- and gamma-terpineol

CAS: 98-55-5 | C10H18O | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 203742500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98-55-5
IUPAC Name2-[(1R)-4-methylcyclohex-3-en-1-yl]propan-2-ol
Molecular FormulaC10H18O
InChI KeyWUOACPNHFRMFPN-VIFPVBQESA-N
SMILESCC1=CC[C@@H](CC1)C(C)(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=94 % (alpha + gamma)
Density(20°C) 0.930 to 0.938 g/ml
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only