Ethyl 3-methyl-2-oxobutyrate, 95%
Ethyl 3-methyl-2-oxobutyrate, 95%
Ethyl 3-methyl-2-oxobutyrate, 95%
Thermo Scientific Chemicals

Ethyl 3-methyl-2-oxobutyrate, 95%

CAS: 20201-24-5 | C7H12O3 | 144.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 203800250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS20201-24-5
IUPAC Nameethyl 3-methyl-2-oxobutanoate
Molecular FormulaC7H12O3
InChI KeyCKTYYUQUWFEUCO-UHFFFAOYSA-N
SMILESCCOC(=O)C(=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4090 to 1.4130 (20°C, 589 nm)
Appearance (Color)Clear light yellow
Infrared spectrumConforms
GC>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only