AminoLink™ Plus Immobilization Trial Kit, 2 mL, 1 Column Kit - FAQs
AminoLink™ Plus Immobilization Trial Kit, 2 mL, 1 Column Kit - FAQs
View additional product information for AminoLink™ Plus Immobilization Kit, 2 mL - FAQs (44894, 20394)
13 product FAQs found
What is the binding capacity of the AminoLink Coupling Resin or aminoLink Plus Resin?
AminoLink Coupling Resin can bind approximately 10 mg of IgG/mL of resin bed, while AminoLink Plus Resin can bind up to 15-20 mg of IgG/mL of resin bed. The upper limit for binding peptides is approximately 1 mg/mL of resin for both support types.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What are the differences between the two supports for the AminoLink Plus Resin?
The AminoLink Plus Resin uses a type of agarose that has better flow properties, which may shorten the time needed for drip-column methods. It is also activated at a higher level, allowing greater ligand to be immobilized.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
My ligand is not soluble in the coupling buffer when using the AminoLink Coupling Resins or Kits. What can I do?
Coupling reactions with AminoLink Resins are compatible with 3-4 M fresh urea or guanidine. (However, old solutions of urea may contain ammonia, which will interfere with ligand immobilization.) Alternatively, dissolve the peptide in 100% DMSO. Add the peptide in DMSO to coupling buffer so that the DMSO does not exceed 20% of the final solution.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How many purifications can be performed using the same AminoLink or AminoLink Plus Column?
Columns can be reused at least 10 times without loss of activity.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How long will the coupling reaction take when using the AminoLink Resins and Kits?
The immobilizatoin reaction takes six hours; the entire procedure may require seven to eight hours. This time can be decreased if the high pH protocol for AminoLink Plus Resin is used.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How can I perform affinity purifications with conjugated AminoLink and AminoLink Plus Affinity Columns?
Equilibrate the prepared column with three to five bed volumes of an appropriate binding buffer. Add 1 mL of sample for each 2 mL bed-volume (serum should be diluted at least 1:1 with binding buffer). Add an additional 200µL of binding buffer to ensure that the entire sample has entered the gel bed. Cap the column bottom and top. Incubate one hour. Wash away non-bound proteins with five to seven bed-volumes of binding buffer or 1 M NaCl. Elute the bound sample by adding small fractions (0.5-1.0 mL) of elution buffer such as Thermo Scientific IgG Elution Buffer (Cat. No. 21004).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How do I determine the degree of conjugation of the immobilized ligand when using the AminoLink Resins and Kits?
Protein samples can be quantified using Thermo Scientific BCA Protein Assay (Cat. No. 23225) or Coomassie Plus Protein Assay (Cat. No. 23236).
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How do AminoLink Resins and Kits work?
Both AminoLink Coupling Resin and AminoLink Plus Coupling Resin consist of a crosslinked beaded agarose that is derivatized to have aldehydes present for coupling. The aldehydes will react with primary amines to form reversible Schiff's bases. After this double bond reacts with sodium cyanoborohydride, the process of reductive amination produces a permanent covalent bond. The aldehyde groups attach directly to the beads without any spacer arms.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Does sodium azide interfere with AminoLink Support coupling?
No, the gel is stored in sodium azide with no loss in activity. Azide is not a primary amine and is rather unreactive.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How do I know whether to use pH 7.2 or pH 10 for coupling my protein to AminoLink Plus Gel?
Coupling at pH 10 generally gives some increase in efficiency but may damage some ligands and requires a more involved procedure. If stability of the ligand is in question or the increase in coupling efficiency is of minor importance, use the pH 7.2 coupling procedure. This procedure has fewer steps and will still give good coupling efficiency.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the difference between AminoLink Coupling Resin and NHS-Activated Agarose?
Both react with primary amines, but AminoLink Coupling Resin is a two-step conjugation. First a Schiff base is formed between the amine and the aldehyde on the resin, which is then reduced to a stable secondary amine with sodium cyanoborohydride, whereas NHS-Activated Agarose utilizes an NHS ester to form an amide bond in a one-step conjugation.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the difference between AminoLink and AminoLink Plus supports?
AminoLink Plus Coupling Resin is activated at a higher level and has higher flow rates than the original AminoLink Coupling Resin, resulting in higher capacity and faster purification. AminoLink Supports can be used to immobilize any molecule with a primary amine.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
How do your AminoLink Coupling Resin and AminoLink Plus Coupling Resin work?
AminoLink and AminoLink Plus Supports are activated with aldehyde groups which will react with primary amines to form Schiff bases, which are reduced to stable, non-reversible secondary amines. Coupling efficiency often exceeds 85% with this support.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.