Cyclophosphamide monohydrate, 97%
Cyclophosphamide monohydrate, 97%
Cyclophosphamide monohydrate, 97%
Thermo Scientific Chemicals

Cyclophosphamide monohydrate, 97%

CAS: 6055-19-2 | C7H17Cl2N2O3P | 279.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 203960250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6055-19-2
IUPAC Name2-[bis(2-chloroethyl)amino]-1,3,2λ⁵-oxazaphosphinan-2-one hydrate
Molecular FormulaC7H17Cl2N2O3P
InChI KeyPWOQRKCAHTVFLB-UHFFFAOYNA-N
SMILESO.ClCCN(CCCl)P1(=O)NCCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Melting point48°C to 53°C
HPLC>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only