Tetraphenylphosphonium bromide, 99%
Tetraphenylphosphonium bromide, 99%
Tetraphenylphosphonium bromide, 99%
Thermo Scientific Chemicals

Tetraphenylphosphonium bromide, 99%

CAS: 2751-90-8 | C24H20BrP | 419.30 g/mol
Quantity:
5 g
25 g
100 g
Packaging:
View all
Plastic bottle
Catalog number 204020250
Price (JPY)
-
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS2751-90-8
IUPAC Nametetraphenylphosphanium bromide
Molecular FormulaC24H20BrP
InChI KeyBRKFQVAOMSWFDU-UHFFFAOYSA-M
SMILES[Br-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point296°C to 300°C
Titration Argentometric>=98.5 %
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
RUO – Research Use Only