Boron trifluoride dimethyl etherate
Boron trifluoride dimethyl etherate
Boron trifluoride dimethyl etherate
Thermo Scientific Chemicals

Boron trifluoride dimethyl etherate

CAS: 353-42-4 | C2H6BF3O | 113.87 g/mol
Have Questions?
Catalog NumberQuantityPackaging
20406050050 mLGlass bottle
Catalog number 204060500
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.30
Save 7.65
Each
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.30
Save 7.65
Each
Chemical Identifiers
CAS353-42-4
IUPAC Namemethoxymethane; trifluoroborane
Molecular FormulaC2H6BF3O
InChI KeyKTPWETRNSUKEME-UHFFFAOYSA-N
SMILESCOC.FB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow to brown
Infrared spectrumConforms
Refractive index1.3070 to 1.3090 (20°C, 589 nm)
Assay>=59 % (BF3)
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only