Pierce™ Mercaptoethylamine-HCl
Thermo Scientific™

Pierce™ Mercaptoethylamine-HCl

Thermo Scientific Pierce 2-Mercaptoethylamine-HCl, commonly abbreviated 2-MEA, is a mild reductant that is often used to selectively reduce hinge-region disulfide자세히 알아보기
Have Questions?
카탈로그 번호수량
204086 x 6 mg
카탈로그 번호 20408
제품 가격(KRW)
287,000
온라인 행사
Ends: 30-Sep-2026
337,000
할인액 50,000 (15%)
Each
수량:
6 x 6 mg
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
287,000
온라인 행사
Ends: 30-Sep-2026
337,000
할인액 50,000 (15%)
Each
Thermo Scientific Pierce 2-Mercaptoethylamine-HCl, commonly abbreviated 2-MEA, is a mild reductant that is often used to selectively reduce hinge-region disulfide bonds in antibody.
For Research Use Only. Not for use in diagnostic procedures.
사양
형태Solid
제품라인Pierce
수량6 x 6 mg
제품 유형Mercaptoethylamine Hydrochloride
Unit SizeEach
구성 및 보관
Store desiccated at 4°C. Product is shipped at ambient temperature.