N,N-Dimethyl-1-naphthylamine, 99%
N,N-Dimethyl-1-naphthylamine, 99%
N,N-Dimethyl-1-naphthylamine, 99%
Acros Organics

N,N-Dimethyl-1-naphthylamine, 99%

CAS: 86-56-6 | C12H13N | 171.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Catalog number 204175000
also known as 20417
HSN 29214590
Price (INR)
377,341.00
Each
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
377,341.00
Each
Chemical Identifiers
CAS86-56-6
IUPAC NameN,N-dimethylnaphthalen-1-amine
Molecular FormulaC12H13N
InChI KeyAJUXDFHPVZQOGF-UHFFFAOYSA-N
SMILESCN(C)C1=C2C=CC=CC2=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.6210 to 1.6230 (20°C, 589 nm)
Appearance (Color)Clear light yellow to brown
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only